Learn More
Medchemexpress LLC 1,4-anthraquinone | 635-12-1 | MFCD00013724 | 95.0% | 208.21 g/mol | C14H8O2 | 1 ML

Supplier: Medchemexpress LLC HYW07724210MM/1ML
1,4-anthraquinone is a small-molecule quinone used in biochemical and pharmacological research. Supplied as a 10 mM solution in DMSO, this product is intended for laboratory research applications and assay use.
- Used in studies of nucleoside transport inhibition.
- Inhibits macromolecule synthesis.
- Induces DNA fragmentation in cellular assays.
- Purity approximately 95.0%.
- Molecular formula C14H8O2, molecular weight 208.21 g/mol.
- Provided as a 10 mM solution in DMSO, 1 mL volume.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.