Learn More
AVANTOR MASTERFLEX (VWR LLC) BarbLock Ultra-Secure Retainer, "" for "" x 3/8"" tubing
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3060400
Encompass
BarbLock assemblies provide uniform 360-degree radial crimping for a leak-free, durable seal. The BarbLock assembly consists of three pieces: the sleeve, collet, and barbed fittings. To install the fitting, slide the sleeve and collet onto the tubing and then insert the barbed fitting into the tubing. Prelock the sleeve and collet by pressing them together until you hear a "click." Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool. Once assembled, the BarbLock assembly can only be unlocked with the removal tool.
Catalog No. NC0125253