Learn More
Medchemexpress LLC 1,4-Benzenediacetic acid | 7325-46-4 | 96.8% | 194.19 | 10 G

Supplier: Medchemexpress LLC HYW00994610G
1,4-Benzenediacetic acid is a chemical compound with the molecular formula C10H10O4 and a molecular weight of 194.19. It is typically supplied as an off-white to light brown solid, with a purity of 96.84% as determined by HPLC. This compound is intended for research use only.
- Molecular formula C10H10O4
- Molecular weight 194.19
- High purity of 96.84% (HPLC)
- Appearance as an off-white to light brown solid
- Stable storage conditions for powder (up to 3 years at -20°C)
- 1H NMR spectrum consistent with its structure
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.