Learn More
Medchemexpress LLC 1,4-Benzenediacetic acid | 7325-46-4 | MFCD00004354 | 96.8% | 194.19 | 50 G

Supplier: Medchemexpress LLC HYW00994650G
1,4-Benzenediacetic acid is a chemical compound with the molecular formula C10H10O4 and a molecular weight of 194.19. Identified as an irritant, this product is intended for research use only and has not been fully validated for medical applications.
- Appearance: Off-white to light brown solid
- Molecular formula: C10H10O4
- Molecular weight: 194.19
- Purity (HPLC): 96.84%
- CAS number: 7325-46-4
- MFCD number: MFCD00004354
- Storage (powder): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.