Learn More
U.S. Pharmacopeia 1,4-Benzoquinone, 106-51-4, MFCD00001591, 200 mg
Supplier:  U.S. Pharmacopeia 1056504
Encompass
Linear Formula: C6H4(=O)2, MW: 108.09, MP: 113-115 °C, Synonym: p-Benzoquinone, Quinone.
Catalog No. NC9121981