Learn More
Medchemexpress LLC 1,4-benzothiazepine, 2,3,4,5-tetrahydro-7-methoxy-4-methyl | 927871-76-9 | MFCD12546447 | 99.8% | 209.31 g·mol⁻¹ | C11H15NOS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1529210MG
Encompass_Preferred
S107 is a small-molecule calstabin 2 modulator used in preclinical research. It is orally available and blood-brain barrier permeable, and stabilizes ryanodine receptor 2 (RyR2) channels by enhancing calstabin 2 binding. This mechanism reduces sarcoplasmic reticulum Ca2+ leak and has been evaluated in studies of cardiac arrhythmia and seizure threshold.
  • Blood-brain barrier permeable.
  • Stabilizes RyR2 channels by enhancing calstabin 2 binding.
  • Reduces sarcoplasmic reticulum Ca2+ leak.
  • Orally bioavailable in preclinical models.
  • High purity solid suitable for research applications.
  • Stable powder with recommended cold storage conditions.
  • Well characterized chemically (formula C11H15NOS; MW 209.31 g·mol⁻¹).
Catalog No. 50-002-87639