Learn More
Medchemexpress LLC 1,4-Bis(diphenylphosphino)butane | 7688-25-7 | 426.47 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00756950G
Encompass_Preferred
1,4-Bis(diphenylphosphino)butane is a biochemical reagent for life science-related research. It serves as a ligand in various reactions, including palladium(0)-catalyzed acylcyanation of arylacetylenes and deprotection of allyloxycarbamates. It also finds use in organometallic chemistry, alkylation, and isomerization reactions.
  • Used in Suzuki, Heck, Negishi, and Sonogashira coupling reactions
  • Acts as a ligand for alkylations and isomerization reactions
  • Essential in organometallic chemistry
Catalog No. 50-004-46589