Learn More
Medchemexpress LLC 1,4-Bis-maleimidobutane | 28537-70-4 | 98.0% | 248.23 | 50 MG

Supplier: Medchemexpress LLC HYW01195750MG
1,4-Bis-maleimidobutane is a biochemical reagent used in various laboratory applications. It is supplied as a white to off-white solid with a purity of 98.0% and a molecular weight of 248.23.
- Used as a biochemical reagent
- High purity of 98.0%
- Solid appearance, white to off-white color
- Molecular weight of 248.23
- Recommended storage conditions for powder and in solvent
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.