Learn More
Medchemexpress LLC 1-(4-(bis(methyl-13C-d3)amino)phenyl)-2-bromoethan-1-one | MFCD06409221 | 98.1% | 250.13 | C813C2H6D6BrNO | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW128417S1MG
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a stable-isotope labeled tracer and internal standard intended for quantitative analytical workflows. Supplied in a 1 mg amount with an accompanying certificate of analysis, it is used for method development, tracer studies, and competition binding experiments in small-scale analytical applications.
  • Stable-isotope labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • High chemical purity suitable for analytical applications.
  • Provided with certificate of analysis for traceability.
  • Small 1 mg size suitable for standards and method development.
Catalog No. 50-002-82579