Learn More
Medchemexpress LLC 1-(4-(bis(Methyl-13C-d3)amino)phenyl)-2-bromoethan-1-one | MFCD06409221 | 98.1% | 250.13 g·mol⁻1 | C8(13C)2H6D6BrNO | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW128417S5MG
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-13C2,d6 is an isotopically labeled reagent designed for analytical applications where precise mass differentiation is required. The molecule carries 13C and deuterium labels to provide a clear mass shift for use as an internal standard or tracer in mass spectrometry, ADME, and metabolism studies.
  • Isotopically labeled (13C2, d6) for mass spectrometry specificity.
  • High reported purity suitable for analytical standards.
  • Solid form with recommended storage for long-term stability.
  • Available in small quantitative packs for trace-level studies.
  • Suitable as an internal standard and tracer in ADME and metabolism assays.
  • Certificate of analysis and structure information available.
Catalog No. 50-002-82959