Learn More
Medchemexpress LLC 1-(4-bromophenyl)-2-[2-(2-thienyl)-4H-triazolo[1,5-a]benzimidazol-4-yl]ethanone | 380645-50-1 | 98.1% | 437.31 g·mol⁻1 | C20H13BrN4OS | 10MG

Supplier: Medchemexpress LLC HY12598010MG
LX-3 is a small-molecule selective activator of the p38 mitogen-activated protein kinase (MAPK) signaling pathway that de-represses EGFP reporter genes silenced by DNA methylation, including TNF, EGR1, LY6K, and ISG20. It is supplied as a high-purity research reagent in pre-measured pack sizes for in vitro studies.
- Selective activator of p38 MAPK signaling.
- De-represses EGFP reporter genes silenced by DNA methylation.
- Solid, off-white to light yellow physical form.
- Molecular formula C20H13BrN4OS, molecular weight 437.31 g·mol⁻¹.
- Purity 98.1% as listed.
- Available in multiple pre-weighed pack sizes (5 mg-200 mg).
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.