Learn More
Medchemexpress LLC 1-(4-chloro-3-methylphenyl)-3-(3,4-difluorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4(5H)-one | 2092923-21-0 | MFCD32174242 | 99.5% | 438.85 | C24H17ClF2N2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10128150MG
Encompass_Preferred
VU 6008667 is a research compound that functions as a selective negative allosteric modulator of the M5 muscarinic acetylcholine receptor, with reported micromolar inhibitory potency and central nervous system penetration suitable for pharmacology studies. It is supplied as a solid for use in in vitro and in vivo experiments.
  • Selective M5 negative allosteric modulator with IC50 = 1.2 μM (human) and 1.6 μM (rat).
  • Demonstrates high central nervous system penetration.
  • High chemical purity (≈99.5%).
  • Supplied as a light yellow solid; soluble in DMSO up to 100 mg/mL.
  • Recommended storage: 4 °C under nitrogen; in solvent: -80 °C (6 months), -20 °C (1 month).
Catalog No. 50-002-13191