Learn More
Medchemexpress LLC 1-(4-chlorophenyl)ethanol | 3391-10-4 | MFCD00041038 | 99.2% | 156.61 g/mol | C8H9ClO | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0744865G
Encompass_Preferred
1-(4-chlorophenyl)ethanol is an aromatic secondary alcohol used as a biochemical reagent and synthetic building block in research applications. It has molecular formula C8H9ClO, molecular weight 156.61 g/mol, CAS 3391-10-4, and is supplied at high purity (HPLC 99.24%). Packaged as 5 g for laboratory use.
  • Aromatic secondary alcohol used as a building block in organic synthesis.
  • High purity suitable for analytical and preparative work (HPLC purity ~99.2%).
  • Supplied in a convenient small-scale package for laboratory use.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-10604