Learn More
Medchemexpress LLC 1-[(4-chlorophenyl)methyl]-5-(trifluoromethyl)-1H-benzimidazol-2-amine | 150493-34-8 | MFCD00920837 | 99.8% | 325.72 | C15H11ClF3N3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10142810MG
Encompass_Preferred
NS-638 is a nonpeptide small molecule L-type calcium channel blocker used in research to inhibit K+-stimulated intracellular Ca2+ elevation. It has reported activity in neuronal assays and is supplied as solid forms and as a DMSO solution for experimental applications.
  • Small nonpeptide calcium channel blocker.
  • Reported IC50 of 3.4 μM for blocking K+-stimulated intracellular Ca2+ elevation.
  • Additional reported activities in neuronal assays (IC50s of ~2.3 and ~4.3 μM in related assays).
  • Molecular formula C15H11ClF3N3 and molecular weight 325.72.
  • High purity (~99.8%) suitable for research use.
  • Available as solid packages and as a 10 mM solution in DMSO for assay preparation.
Catalog No. 50-002-49280