Learn More
Medchemexpress LLC HY-N7117 100mg Medchemexpress, 1,4-Cineole CAS:470-67-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7117100MG
Encompass_Preferred
Medchemexpress, HY-N7117 100mg 1,4-Cineole CAS:470-67-7 1,4-Cineole is a widely distributed, natural, oxygenated monoterpene. 1,4-Cineole, present in eucalyptus oil, activates both human TRPM8 and human TRPA1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0505