Learn More
Cayman Chemical 1 4-D-XylobIose 10mg
SDP
Supplier:  Cayman Chemical 3403110 MG
Encompass_Preferred
A disaccharide; a degradation product of hemicellulose; has been found in T. reesei endoxylanase-treated birch kraft pulp
Catalog No. 50-294-7174