Learn More
Medchemexpress LLC 1,4-diazoniabicyclo[2.2.2]octane, 1,4-disulfino-, bis(inner salt) | 119752-83-9 | MFCD22377802 | 97.0% | 240.31 | C6H12N2O4S2 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW033049
Encompass_Preferred
DABCO-Bis(sulfur dioxide) is a bench-stable crystalline adduct that acts as a convenient, solid-source surrogate for sulfur dioxide in organic synthesis. It enables controlled SO2 transfer for the preparation of sulfonamides, sulfones, and sulfinate intermediates, simplifying handling compared with gaseous SO2.
  • Bench-stable solid for safer handling and storage.
  • Provides controlled sulfur dioxide transfer in reactions.
  • Suitable for preparation of sulfonamides and sulfones.
  • Reported purity 97.0%.
  • Molecular weight 240.31; formula C6H12N2O4S2.
  • Available in multiple laboratory-scale pack sizes.
Catalog No. 50-003-02777