Learn More
Medchemexpress LLC 1,4-Dibromobenzene-d4 (p-dibromobenzene-d4) | 4165-56-4 | 99.9% | C6D4Br2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY76025S10G
Encompass_Preferred
1,4-Dibromobenzene-d4 is a deuterium labeled 1,4-dibromobenzene. The unlabeled compound is used as a fumigant and a chemical intermediate for pharmaceuticals. Inhalation exposure can cause dizziness and choking, while contact with skin or eyes may produce inflammation and burning.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-34193