Learn More
Cayman Chemical 1 4-Dicaffeoylquinic acid 5MG
SDP
Supplier:  Cayman Chemical 338425 MG
Encompass_Preferred
A polyphenol with diverse biological activities; scavenges DPPH radicals; inhibits XO (IC50 = 7.36 µM); decreases protein levels of tyrosinase and MITF in B16/F1 murine melanocytes, inhibits melanogenesis in the same cells, and decreases tyrosinase activity in cell lysates at 25 µM; increases biliary flow in rats
Catalog No. 50-294-6417