Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Diiodobenzene | 624-38-4 | 329.91 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW007412

Encompass_Preferred

1,4-Diiodobenzene is a biochemical reagent designed for life science research. It can be utilized as a biological material or an organic compound in various scientific applications.

  • Suitable for life science-related research
  • Can be used as a biological material
  • Can be used as an organic compound
  • Appearance: Solid
  • Color: Light yellow to brown
  • Formula: C6H4I2
  • For research use only

Catalog No. 50-004-49178


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.