Learn More
Medchemexpress LLC 1-(4-dimethylphosphoryl-2-methyl-6-propan-2-ylphenyl)-6-fluoro-7-(2-fluoro-6-hydroxyphenyl)-4-[(2S)-2-methyl-4-prop-2-enoylpiperazin-1-yl]pyrido[2,3-d]py. | 2649004-88-4 | 99.9% | 635.64 g/mol | C33H36F2N5O4P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1424605MG
Encompass_Preferred
KRAS G12C inhibitor 28 is a research-grade small-molecule inhibitor selective for the KRAS G12C mutant. It shows potent in vitro activity (IC50 57 nM) and has reported antitumor effects in preclinical examples (WO2021113595A1). The compound is supplied as a high-purity solid for biochemical and cellular research applications.
  • Potent KRAS G12C inhibition (IC50 57 nM).
  • Demonstrated antitumor effects in preclinical examples.
  • High chemical purity suitable for biochemical and cellular assays.
  • Available in multiple small-mass sizes for research use.
  • Molecular weight 635.64 and formula C33H36F2N5O4P for formulation planning.
Catalog No. 50-002-21268