Learn More
Medchemexpress LLC 1-((4-fluorophenyl)thio)isoquinoline | 864863-72-9 | 99.9% | 305.31 | C17H12FN5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10038210MM 1ML
Encompass_Preferred
FPTQ is a potent mGluR1 antagonist with low-nanomolar activity (human IC50 ≈ 6 nM; mouse IC50 ≈ 1.4 nM). It has reported antioxidant and anti-inflammatory effects in vitro and in vivo, and is supplied for research as a solid or as a 10 mM solution in DMSO. High purity and recommended storage conditions support reliable experimental use.
  • Potent mGluR1 antagonist with low-nanomolar potency.
  • Demonstrated antioxidant and anti-inflammatory activity in biological studies.
  • Available as solid or 10 mM solution in DMSO for experimental convenience.
  • High purity for reproducible results.
  • Recommended storage conditions provided to maintain stability.
Catalog No. 50-002-14592