Learn More
Medchemexpress LLC 1-(4-methoxyphenyl)-N-[(4-propan-2-ylphenyl)methyl]benzimidazol-5-amine | 489416-12-8 | 99.9% | 371.47 g/mol | C24H25N3O | 100 MG

Supplier: Medchemexpress LLC HY101441100MG
ST-193 is a small-molecule research compound that functions as a potent, broad-spectrum arenavirus inhibitor. It shows low-nanomolar activity against multiple pathogenic arenaviruses and is used for in vitro and preclinical research. CAS 489416-12-8.
- Potent broad-spectrum arenavirus inhibitor with low-nanomolar IC50 values.
- Soluble in DMSO (100 mg/mL) and in selected formulation vehicles at ≥ 2.5 mg/mL.
- Molecular formula C24H25N3O; molecular weight 371.47 g/mol.
- Storage: powder -20°C (stable 3 years) or 4°C (stable 2 years); in solvent -80°C (2 years).
- Intended for in vitro and preclinical research targeting arenavirus infection pathways.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.