Learn More
Cayman Chemical 1 4-PBITdIhydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 8151010 MG
Encompass_Preferred
1,4-PBIT is a potent inhibitor of purified human iNOS and nNOS with Ki values of 7.4 and 16 nM, respectively, compared to a Ki of 360 nM for human eNOS.{1292} Its inhibition in whole cells is greatly diminished, presumably to poor membrane permeability.
Catalog No. 50-289-4830