Learn More
Medchemexpress LLC 1,4-Pentanediamine, N4-[7-chloro-2-[(1E)-2-(2-chlorophenyl)ethenyl]-4-quinolinyl]-N1,N1-diethyl | 529507-84-4 | 99.7% | 1 MG

Supplier: Medchemexpress LLC HY1199101MG
(E)-Aminoquinol is a small-molecule GFRα-1 agonist used in preclinical studies to mimic GDNF neurotrophic effects and induce Ret autophosphorylation in neuronal cells. It is supplied as a high-purity solid for research applications.
- Purity 99.7%
- Molecular weight 456.45 g/mol
- Chemical formula C26H31Cl2N3
- Appearance white to off-white solid
- Storage 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month
- Available pack sizes 1 mg, 5 mg, 10 mg, 50 mg
- Target GFRα-1, used in Parkinson's disease research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.