Learn More
Waters Corp Pinch Valve, 1/4 in., for QDa Systems, MS System Type
Supplier:  Waters Corp 700010241
Encompass_Preferred
This is a replacement pinch valve for QDa systems with a serial prefix KAB. The valve is located outside the QDa unit.
  • Product type: Spare parts kits
  • Type: Valve
  • System type: MS
  • Replacement schedule: As required
  • Designed for QDa systems with a serial prefix KAB
  • Situated outside the QDa
  • 1/4 in. size
Catalog No. 50-239-2930