Learn More
Cayman Chemical 1 4dIdeoxy1 4ImInoDArabI 10mg
SDP
Supplier:  Cayman Chemical 2093910 MG
Encompass_Preferred
An inhibitor of glycogen phosphorylase; inhibits glycogenolysis in isolated liver cells (IC50 = 1.0 µM) and in homogenates of cerebral cortex and cerebellum (IC50s = 463 and 383 nM, respectively)
Catalog No. 50-298-3268