Learn More
BRANSON ULTRASONICS CORPORATION (3P) 1/4IN MICROTIP 500/550/250/450
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) 101148070
Encompass_Preferred
Branson Ultrasonics Microtips for 200 and 400 Watt Sonifiers, 1/4 in. tapered tip; with coupler and lower portion, Amplitude Range: 59.5 to 247um, Diameter Tip: 0.25 in., 0.65cm, Intensity: High, Tip Style: Tapered, Capacity: 10 to 50mL
Catalog No. NC3721568