Learn More
Chemglass Life Sciences Adapter, TFE Compression, 1/4" OD to 3/4" NPT Male
SDP
Supplier:  Chemglass Life Sciences CG197181J
Encompass_Preferred
Adapter, TFE Compression, 1/4" OD to 3/4" NPT Male. Designed for use with Chemglass's FEP tubing. Fittings are made entirely of chemically inert corrosion resistant PTFE. Fittings: Machined from virgin Polytetrafluoroethylene. Ferrules: Ferrules are machined from virgin Polytetrafluoroethylene. The Gripper in the two piece Ferrule-Gripper assemblies are manufactured from PVDF. Nuts are molded PVDF.
Catalog No. 50-153-1353