Learn More
Medchemexpress LLC 1,5-dimethyl-4-(methylamino)-2-phenylpyrazol-3-one | 519-98-2 | MFCD00447796 | 99.8% | 217.27 g·mol⁻¹ | C12H15N3O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1357315MG
Encompass_Preferred
4-Methylamino antipyrine is an active metabolite of metamizole used as a research reference standard. It exhibits analgesic and antipyretic activity and has been reported to weakly inhibit cyclooxygenase (COX). Supplied as a high-purity solid suitable for analytical, metabolic, and pharmacological studies.
  • High purity for analytical use.
  • Analgesic and antipyretic pharmacological profile.
  • Suitable as a reference standard in metabolism studies.
  • Available in multiple pack sizes for small-scale research.
  • Molecular formula C12H15N3O; molecular weight 217.27 g·mol⁻¹.
Catalog No. 50-002-22357