Learn More
Medchemexpress LLC 1-[5-ethyl-2-hydroxy-4-[6-methyl-6-(2H-tetrazol-5-yl)heptoxy]phenyl]ethanone | 117690-79-6 | MFCD00876422 | 98.0% | 360.45 g/mol | C19H28N4O3 | 10 MG

Supplier: Medchemexpress LLC HY1574410MG
LY255283 is a small-molecule antagonist of the leukotriene B4 (BLT2) receptor used as a research tool to probe BLT2-mediated signaling and inflammatory responses. It is reported to displace [3H]LTB4 with an IC50 of approximately 100 nM in guinea pig lung membrane assays and is supplied as a solid research-grade powder.
- Small-molecule BLT2 (LTB4) receptor antagonist for research use.
- Reported IC50 ~100 nM for [3H]LTB4 binding to guinea pig lung membranes.
- Chemical formula C19H28N4O3; molecular weight 360.45 g/mol.
- Purity typically ≥98% (HPLC), supplied as a solid powder.
- Suitable for biochemical and cellular assays to study leukotriene signaling and inflammation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.