Learn More
Andwin Scientific 1.5 ML CONICAL SAF-T-SEAL SCRE
SDP
Supplier:  Andwin Scientific 14158707
Encompass_Preferred
The 0.5 mL conical screw cap tube is an ideal laboratory apparatus for centrifugation up to 20000 x g, vapor-phase liquid nitrogen storage, -80C freezers, boiling, and autoclaving. The tube's external threads prevent sample loss and seal failure. This product is autoclavable and free of RNase, DNase, DNA, and pyrogen. It comes in a pack of 500 tubes with amber colored caps.
Catalog No. 50-819-725