Learn More
Andwin Scientific 1.5 ML SEAL-RITE MICROCENTRIF
SDP
Supplier:  Andwin Scientific 16155507
Encompass_Preferred
The 1.5 ml microcentrifuge tube without cap is an ideal laboratory apparatus for column fractions, vacuum concentration, and other procedures where caps interfere. This product also eliminates the time need to cut caps off tubes. It comes in a bag of 1000 natural color polypropylene tubes.
Catalog No. 50-819-855