Learn More
Andwin Scientific 1.5 ML SELF-STANDING SAF-T-SEA
SDP
Supplier:  Andwin Scientific 14159707
Encompass
The 0.5 mL self-standing screw cap tube is an ideal laboratory apparatus for centrifugation up to 20000 x g, vapor-phase liquid nitrogen storage, -80C freezers, boiling, and autoclaving. The tube's external threads prevent sample loss and seal failure. This product is autoclavable and free of RNase, DNase, DNA, and pyrogen. It comes in a pack of 500 tubes with amber colored caps.
Catalog No. 50-819-744