Learn More
Medchemexpress LLC 1-[5-(thiophen-2-ylmethoxy)-1H-indol-3-yl]propan-2-amine;hydrochloride | 160521-72-2 | MFCD01321066 | 99.8% | 322.85 | C16H19ClN2OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10136910MG
Encompass_Preferred
BW-723C86 is a research-grade selective 5-HT2B receptor agonist provided in solid and DMSO solution formats for in vitro and in vivo pharmacology studies. It exhibits high purity and has documented solubility and storage recommendations to support experimental reproducibility. Use only as directed for research purposes and follow safety and handling instructions from the supplier's safety documentation.
  • Selective 5-HT2B receptor agonist.
  • High purity suitable for pharmacological assays.
  • Available as solid or ready-to-use DMSO solution.
  • High solubility in DMSO (≈100 mg/mL) for easy formulation.
  • In vivo formulation protocols provided to aid dosing.
  • Storage recommendations provided to preserve stability.
  • Intended for research use only.
Catalog No. 50-002-11646