Learn More
AirSea USA 1.5L Polyporopylene Plastic Biojar
Supplier:  AirSea USA 490
Encompass_Preferred
1.5L Biojar with 120mm yellow screw cap - capable of withstanding a pressure of 95kPa and suitable for shipping infectious substances inside inner receptacles. Biojar is fully autoclavable. This biojar performs to the packing instruction 620. We have a range of absorbent materials and cushioning here.
Catalog No. 50-153-4814