Learn More
CELLTREAT Scientific Products 1.5mL Micro Centrifuge Tube, Assorted Color, Non-sterile
SDP
Supplier:  CELLTREAT Scientific Products 229444A
Encompass_Preferred
All tubes have a frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handColor tubes allow for quick sample identification, or to customize workflows
Catalog No. 50-001-44330