Learn More
Polycarbin 1.5 mL Microcentrifuge Tubes CS5000 (Non-sterile)
SDP
Supplier:  Polycarbin PC020301
Encompass_Preferred
Polycarbin's 1.5 mL microcentrifuge tube is made using 20% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint. Microcentrifuge tubes are certified DNase, RNase, and Pyrogen free. Non-sterile (autoclavable). Max RCF: 54,234 (21,000 RPM). 500 tubes/bag, 10 bags/case, 5000 tubes/case. Manufactured in an ISO 9001 compliant facility.
Catalog No. NC2953702