Learn More
NEST SCIENTIFIC USA 1.5ml Microcentrifuge Tube, Clear, Conical, Lock Cap, 5000/cs
Supplier:  NEST SCIENTIFIC USA 615001
Encompass_Preferred
Nest microcentrifuge tubes are molded from USP grade VI polypropylene. Made using uses diamond polished molds for superior tolerances. Excellent sealing and low-retention perfomance without PFAS additives. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).
Catalog No. NC3429298