Learn More
Chemglass Life Sciences Chemglass 1.5 mL Self-Standing Vial, External Thread, Natural Cap, Sterile (50/pack, Nest)
SDP
Supplier:  Chemglass Life Sciences CGN4764003N
Encompass_Preferred
1.5 mL self-standing vials with external thread and natural-color sealing caps, supplied sterile in nest packaging. Designed for secure sample storage and handling in laboratory applications; manufactured in a controlled clean workshop and sterilized by electron beam to a sterility assurance level of 10^-6.
  • Capacity: 1.5 mL.
  • Self-standing design for upright storage.
  • External thread with sealing ring for leak resistance.
  • Natural-color caps provided.
  • Supplied sterile (electron beam sterilization, SAL = 10^-6).
  • Packaged in nest format: 50 per pack, 500 per box, 2,000 per case.
  • Manufactured in a 100,000-level clean workshop; DNAse-free, RNAse-free, pyrogen-free, endotoxin-free.
Catalog No. 50-273-3360