Learn More
Chemglass Life Sciences 1.5 mL Self-Standing Vial - External Thread, Hinged Cap with Sealing Ring; Natural Cap; Sterile; 50/Pack
SDP
Supplier:  Chemglass Life Sciences CGN4764013N
Encompass_Preferred
These sterile 1.5 mL self-standing vials are designed for secure storage and handling of low-volume samples. The external thread and hinged cap with sealing ring provide a reliable closure, while nested sterile packaging supports clean workflows and high-throughput use.
  • 1.5 mL capacity for small-volume samples.
  • Self-standing, round-bottom design for stability.
  • External thread with hinged cap and sealing ring for secure closure.
  • Natural color cap to reduce light transmission.
  • Sterile; electron beam sterilized to SAL 10^-6.
  • Packaged 50 per pack, 500 per box, 2,000 per case.
  • Manufactured in a 100,000-level clean workshop.
  • DNAse-free, RNAse-free, pyrogen-free, endotoxin-free.
Catalog No. 50-273-3356