Learn More
Medchemexpress LLC 1,7-Bis(4-hydroxyphenyl)-1,4,6-heptatrien-3-one | 149732-52-5 | 292.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN119025MG
Encompass_Preferred
1,7-Bis(4-hydroxyphenyl)-1,4,6-heptatrien-3-one is a potent nucleocapsid (N) protein inhibitor isolated from *Curcuma kwangsiensis*. It demonstrates excellent antiviral activity against HCoV-OC43 and SARS-CoV-2.
  • Inhibits nucleocapsid (N) protein
  • Exhibits antiviral activity against HCoV-OC43 and SARS-CoV-2
  • Active against various cell lines (KB, PC-12, Vero)
Catalog No. 50-004-14623