Learn More
Medchemexpress LLC 1,7-dimethyluric acid | 33868-03-0 | MFCD00005713 | 99.8% | 196.16 | C7H8N4O3 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1260611MG
Encompass_Preferred
1,7-Dimethyluric acid (CAS 33868-03-0) is a small-molecule metabolite of caffeine commonly used as an analytical standard and research reagent. It is a white to off-white solid with a molecular weight of 196.16 g/mol and reported high purity, suitable for metabolic studies, analytical method development, and reference assays.
  • Small-molecule caffeine metabolite for metabolic research.
  • High purity: 99.8%.
  • White to off-white solid suitable for analytical handling.
  • Used as an analytical standard and reference in assays.
  • Recommended storage under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Applicable to biochemical and analytical workflows.
Catalog No. 50-002-34959