Learn More
Waters Corp Tubing, PTFE, 1/8 in. OD, 0.009 in. ID, 10 ft.
Supplier:  Waters Corp WAT088431
Encompass_Preferred
This tubing is designed for liquid chromatography (LC) applications. It features precise outer and inner diameters to meet instrument requirements and is engineered for optimal performance when used with compatible fittings.
  • Designed to provide leak-free connections
  • Ensures low dead volume
  • Minimizes band spreading
  • Components must be clean and designed to work together for optimal performance
Catalog No. 50-793-767