Learn More
Chemglass Life Sciences Chemglass Excelon PVC standard wall lab tubing, 1/8 in ID x 3/16 in OD, 50 ft roll (CG-730-003)
SDP
Supplier:  Chemglass Life Sciences CG730003
Encompass_Preferred
Chemglass Excelon standard wall PVC tubing is a flexible, general-purpose laboratory tubing with a smooth inner bore to reduce buildup and simplify changeovers. It is suitable for fluid transfer, pump connections, and general lab use, and is supplied on a 50 ft roll for convenient installation.
  • Standard wall PVC tubing for laboratory fluid transfer.
  • Inner diameter 1/8 in, outer diameter 3/16 in, wall thickness 1/32 in.
  • Sold on a 50 ft roll for extended runs and replacement.
  • Smooth inner bore reduces buildup and simplifies changeovers.
  • Flexible and compatible with peristaltic pumps and general connections.
Catalog No. NC2323631