Learn More
Ace Glass, Inc. PTFE 1/8in tube compression fitting to 1/8in male NPT
SDP
Supplier:  Ace Glass, Inc. 1270904
Encompass
1/8 PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-669