Learn More
Medchemexpress LLC 1 9-Bis-boc-1 5 9-tr 5g
SDP
Supplier:  Medchemexpress LLC HYW0921765G
Encompass_Preferred
1 9-Bis-Boc-1 5 9-triazanonane is a linker containing two Boc-protected amino groups The Boc groups can be deprotected under mild acidic conditions to form the free amines
Catalog No. 50-002-06594