Learn More
Medchemexpress LLC 1,9-Diazaspiro[5.5]undecan-2-one, 1-(3,4-difluorophenyl)-9-[6-(2,2,2-trifluoroethoxy)-4-pyrimidinyl | 2761767-18-2 | 98.8% | 456.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY168498
Encompass_Preferred
GJG057 is a selective, orally active inhibitor for leukotriene C4 synthase (LTC4S) with an IC50 of 44 nM. This compound exhibits anti-inflammatory activity in mouse skin inflammation and asthma models.
  • Selective, orally active inhibitor for leukotriene C4 synthase (LTC4S) with an IC50 of 44 nM.
  • Exhibits anti-inflammatory activity in mouse skin inflammation model and mouse asthma model.
  • For research use only.
  • Appearance: Solid, Off-white to light brown.
Catalog No. 50-003-94049