Learn More
Medchemexpress LLC 1-[(9H-Fluoren-2-yl)methyl]-1-cycloheptyl-3-mesitylurea | 138046-43-2 | MFCD00905424 | >98.0% | 452.63 g·mol⁻¹ | C31H36N2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10739625MG
Encompass_Preferred
YM-750 is a research-grade acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor with reported in vitro potency (IC50 = 0.18 μM). It is used in biochemical and cellular studies to probe cholesterol esterification and lipid metabolism. Key chemical identifiers include CAS 138046-43-2, formula C31H36N2O, and molecular weight 452.63 g·mol⁻¹.
  • Potent ACAT inhibitor (IC50 0.18 μM).
  • Suitable for biochemical and cellular assays.
  • High purity suitable for research (>98% by HPLC).
  • Available as solids and as a DMSO solution in multiple package sizes.
  • Consistent chemical identity: CAS 138046-43-2 and formula C31H36N2O.
Catalog No. 50-002-39905