Learn More
Medchemexpress LLC 1-(9H-Fluoren-9-yl)-4,7,10-trimethyl-3,6,9-trioxo-2-oxa-4,7,10-triazadodecan-12-oic acid | 2749824-37-9 | 98.5% | 453.49 | 50 MG
Supplier: Medchemexpress LLC HY16416650MG
Fmoc-Sar-Sar-Sar-OH is a small molecule polypeptide composed of four sarcosine residues, with the N-terminus of the peptide chain protected by Fmoc. It can be used for peptide synthesis or the synthesis of drug-peptide ligand conjugates.
- Used for peptide synthesis
- Used for synthesis of drug-peptide ligand conjugates